Pelitinib (EKB-569) 98%

Supplier: Ambeed
Warning

Synonyms: (E)-N-(4-((3-Chloro-4-fluorophenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide, (E)-N-(4-(3-chloro-4-fluorophenylamino)-3-cyano-7-ethoxyquinolin-6-yl)-4-(dimethylamino)but-2-enamide, EKB 569, Pelitinib, (2E)-N-[4-[(3-Chloro-4-fluorophenyl)amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-dimethylamino-2-butenamide

A111899-1MG A111899-5MG
76718-114EA 10.61 USD
76718-114 76718-116
Pelitinib (EKB-569) 98%
Pelitinib (EKB-569)

Precaución: for Research Use Only

Formula: C₂₄H₂₃ClFN₅O₂
MW: 467.92 g/mol
MDL Number: MFCD09837868
CAS Number: 257933-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR