2-Chloro-4'-nitroacetanilide 95%

Supplier: Ambeed
Danger

A111225-1G A111225-250MG A111225-5G
76718-094EA 18 USD
76718-094 76718-096 76718-098
2-Chloro-4'-nitroacetanilide 95%
2-Chloro-4'-nitroacetanilide

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₃
MDL Number: MFCD00031337
CAS Number: 17329-87-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR