Dicyclohexylamine (2S,3S)-2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate 95%
Supplier: Ambeed
|
Warning
|
A111095-250MG
A111095-5G
76718-090EA
8.49
USD
76718-090
76718-092
Dicyclohexylamine (2S,3S)-2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate 95%
Dicyclohexylamine (2S,3S)-2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate
Precaución: for Research Use Only
Formula:
C₂₁H₄₀N₂O₅ MW: 400.55 g/mol |
MDL Number:
MFCD27920593 CAS Number: 1464025-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...