Octenidine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride

A110333-100G A110333-10G A110333-1G A110333-1MG A110333-250MG A110333-25G A110333-50MG A110333-5G
76718-054EA 481 USD
76718-054 76718-056 76718-058 76718-060 76718-062 76718-064 76718-066 76718-068
Octenidine dihydrochloride 98%
Octenidine dihydrochloride

Precaución: for Research Use Only

Formula: C₃₆H₆₂N₄
MW: 623.84 g/mol
MDL Number: MFCD01938808
CAS Number: 70775-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR