4-(Chloromethyl)-N-(4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide 95%

Supplier: Ambeed
Warning

A109617-250MG A109617-5G
76718-022EA 85.94 USD
76718-022 77666-598
4-(Chloromethyl)-N-(4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide 95%
4-(Chloromethyl)-N-(4-methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClN₅O
MW: 429.90 g/mol
MDL Number: MFCD09841001
CAS Number: 404844-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR