Ethyl 3-oxo-3-(m-tolyl)propanoate 95%
Supplier: Ambeed
|
Warning
|
A109045-10G
A109045-1G
A109045-250MG
A109045-5G
76717-980EA
260.99
USD
76717-980
76717-982
76717-984
76717-986
Ethyl 3-oxo-3-(m-tolyl)propanoate 95%
Ethyl 3-oxo-3-(m-tolyl)propanoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₄O₃ MW: 206.24 g/mol |
MDL Number:
MFCD03424816 CAS Number: 33166-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...