Pranlukast 98%

Supplier: Ambeed
Danger

Synonyms: N-[4-Oxo-2-(1H-tetrazol-5-yl)-4H-1-benzopyran-8-yl]-4(4-phenylbutoxy)benzamide, ONO 1078, N-(4-Oxo-2-(1H-tetrazol-5-yl)-4H-chromen-8-yl)-4-(4-phenylbutoxy)benzamide

A108288-100MG A108288-1G A108288-5G
76717-948EA 6 USD
76717-948 76717-950 77878-819
Pranlukast 98%
Pranlukast

Precaución: for Research Use Only

Formula: C₂₇H₂₃N₅O₄
MDL Number: MFCD00864631
CAS Number: 103177-37-3
UN: 1325
ADR: 4.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR