Phenyl phosphate disodium salt 98%

Supplier: Ambeed
Warning

A108118-100G A108118-1G A108118-25G A108118-500G A108118-5G
76717-918EA 191 USD
76717-918 76717-920 76717-922 76717-924 76717-926
Phenyl phosphate disodium salt 98%
Phenyl phosphate disodium salt

Precaución: for Research Use Only

Formula: C₆H₅Na₂O₄P·xH₂O
MDL Number: MFCD00002133
CAS Number: 3279-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR