Sodium dihydrogen phosphate dihydrate 99%

Supplier: Ambeed
Warning

A107451-1000G A107451-500G A107451-100G
76717-908EA 15 USD
76717-908 76717-910 77599-556
Sodium dihydrogen phosphate dihydrate 99%
Sodium dihydrogen phosphate dihydrate

Precaución: for Research Use Only

Formula: NaH₂PO₄·2H₂O
MDL Number: MFCD00149209
CAS Number: 13472-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR