Dansyl hydrazide 97%

Supplier: Ambeed
Danger

Synonyms: Dansyl hydrazine, 5-Dimethylaminonaphthalene-1-sulphonyl hydrazine, 5-(Dimethylamino)naphthalene-1-sulphonohydrazide

A107446-250MG A107446-5G
76717-904EA 26 USD
76717-904 76717-906
Dansyl hydrazide 97%
Dansyl hydrazide

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₂S
MDL Number: MFCD00003986
CAS Number: 33008-06-9
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR