Sodium myristate 98%

Supplier: Ambeed
Warning

Synonyms: Myristic Acid Sodium Salt

A107107-100G A107107-25G A107107-500G A107107-5G
76717-886EA 127.31 USD
76717-886 76717-888 76717-890 76717-892
Sodium myristate 98%
Sodium myristate

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₂
MW: 250.36 g/mol
MDL Number: MFCD00042653
CAS Number: 822-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR