Potassium 2-(methylsulfonyl)phenyltrifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(2-(methylsulphonyl)phenyl)borate, Potassium (2-methylsulphonylphenyl)trifluoroborate

A106745-1G A106745-250MG A106745-5G
76717-874EA 83.82 USD
76717-874 76717-876 76717-878
Potassium 2-(methylsulfonyl)phenyltrifluoroborate 98%
Potassium 2-(methylsulphonyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO₂S
MW: 262.1000 g/mol
MDL Number: MFCD04115760
CAS Number: 850623-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR