2-Cyano-N-ethylacetamide 98%

Supplier: Ambeed
Warning

A106635-100G A106635-25G A106635-5G
76717-860EA 367 USD
76717-860 76717-862 76717-864
2-Cyano-N-ethylacetamide 98%
2-Cyano-N-ethylacetamide

Precaución: for Research Use Only

Formula: C₅H₈N₂O
MDL Number: MFCD00118415
CAS Number: 15029-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR