Purvalanol A 98%

Supplier: Ambeed
Warning

A106338-10MG A106338-1MG A106338-2MG A106338-50MG A106338-5MG
76717-850EA 60 USD
76717-850 76717-852 76717-854 76717-856 76717-858
Purvalanol A 98%
Purvalanol A

Precaución: for Research Use Only

Formula: C₁₉H₂₅N₆OCl
MDL Number: MFCD02179211
CAS Number: 212844-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR