Methyl 3-(trimethylsilyl)-4-pentenoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(trimethylsilyl)pent-4-enoate

A106002-100G A106002-1G
76717-838EA 498.63 USD
76717-838 76717-840
Methyl 3-(trimethylsilyl)-4-pentenoate 97%
Methyl 3-(trimethylsilyl)-4-pentenoate

Precaución: for Research Use Only

Formula: C₉H₁₈O₂Si
MW: 186.324 g/mol
MDL Number: MFCD01863734
CAS Number: 185411-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR