tert-Butyl-4-formylbenzylcarbamate 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Aminomethyl)benzaldehyde, N-BOC protected, tert-Butyl 4-formylbenzylcarbamate

A105737-100G A105737-10G A105737-1G A105737-250MG A105737-25G A105737-5G
76717-812EA 1176 USD
76717-812 76717-814 76717-816 76717-818 76717-820 76717-822
tert-Butyl-4-formylbenzylcarbamate 97%
tert-Butyl-4-formylbenzylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.2800 g/mol
MDL Number: MFCD06659090
CAS Number: 156866-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR