Propafenone hydrochloride 99%

Supplier: Ambeed
Warning

Synonyms: 1-(2-(2-Hydroxy-3-(propylamino)propoxy)phenyl)-3-phenylpropan-1-one hydrochloride, Propafenone HCl

A105689-10G A105689-1G A105689-1MG A105689-250MG A105689-25G A105689-5G A105689-100G
76717-800EA 83 USD
76717-800 76717-802 76717-804 76717-806 76717-808 76717-810 77666-548
Propafenone hydrochloride 99%
Propafenone hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₇NO₃·HCl
MW: 377.9 g/mol
MDL Number: MFCD00079243
CAS Number: 34183-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR