Potassium Butyltrifluoroborate 97%

Supplier: Ambeed
Danger

Synonyms: Potassium butyltrifluoroborate

A105128-1G A105128-250MG A105128-5G A105128-25G
76717-748EA 8.49 USD
76717-748 76717-750 76717-752 77570-306
Potassium Butyltrifluoroborate 97%
Potassium Butyltrifluoroborate

Precaución: for Research Use Only

Formula: C₄H₉BF₃K
MW: 164.02 g/mol
MDL Number: MFCD08276805
CAS Number: 444343-55-9
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR