5-Bromoquinazolin-6-ylthiourea 97%

Supplier: Ambeed
Danger

Synonyms: 1-(5-Bromoquinoxalin-6-yl)thiourea

A104889-100MG A104889-1G A104889-250MG A104889-5G
76717-738EA 35.01 USD
76717-738 76717-740 76717-742 76717-744
5-Bromoquinazolin-6-ylthiourea 97%
5-Bromoquinazolin-6-ylthiourea

Precaución: for Research Use Only

Formula: C₉H₇BrN₄S
MW: 283.147 g/mol
MDL Number: MFCD08445609
CAS Number: 842138-74-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR