Octyl-3,4,5-trihydroxybenzoate 97%

Supplier: Ambeed
Warning

A104868-100G A104868-25G A104868-5G A104868-1G A104868-10MG A104868-500G
76717-732EA 70 USD
76717-732 76717-734 76717-736 77570-288 77599-536 77666-534
Octyl-3,4,5-trihydroxybenzoate 97%
Octyl-3,4,5-trihydroxybenzoate

Precaución: for Research Use Only

Formula: C₁₅H₂₂O₅
MDL Number: MFCD00002197
CAS Number: 1034-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR