(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
((1,3-Dioxolan-2-yl)methyl)triphenylphosphonium bromide
A104641-100G
A104641-10G
A104641-25G
A104641-500G
A104641-5G
76717-706EA
26
USD
76717-706
76717-708
76717-710
76717-712
76717-714
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide 97%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₂₂H₂₂BrO₂P MW: 429.30 g/mol |
MDL Number:
MFCD00011966 CAS Number: 52509-14-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...