Phenoxyacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenoxyacetic acid

A103184-1000G A103184-100G A103184-500G
76717-672EA 51.99 USD
76717-672 76717-674 76717-676
Phenoxyacetic acid 98%
Phenoxyacetic acid

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00004296
CAS Number: 122-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR