Sophoridine 96%

Supplier: Ambeed
Danger

Synonyms: (7aR,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, (5beta)-Matridin-15-one, (41S,7aR,13aR,13bR)-Dodecahydro-1H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10(41H)-one

A102794-1G A102794-25MG A102794-5G A102794-1MG A102794-25G
76717-668EA 21.22 USD
76717-668 76717-670 77599-500 77666-482 77666-484
Sophoridine 96%
Sophoridine

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂O
MW: 248.37 g/mol
MDL Number: MFCD00238686
CAS Number: 6882-68-4
UN: 1544
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR