Silver methanesulfonate 99%
Supplier: Ambeed
|
Warning
|
A102752-100G
A102752-1G
A102752-25G
A102752-5G
76717-660EA
310.85
USD
76717-660
76717-662
76717-664
76717-666
Silver methanesulfonate 99%
Silver methanesulphonate
Precaución: for Research Use Only
Formula:
CH₃AgO₃S MW: 202.97 g/mol |
MDL Number:
MFCD00064795 CAS Number: 2386-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...