Methyl D-homoalaninate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-D-2-Abu-OMe·HCl, (R)-Methyl 2-aminobutanoate hydrochloride

A102572-100G A102572-10G A102572-1G A102572-25G A102572-5G
76717-642EA 246.13 USD
76717-642 76717-644 76717-646 76717-648 76717-650
Methyl D-homoalaninate hydrochloride 98%
Methyl D-homoalaninate hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂·ClH
MW: 153.607 g/mol
MDL Number: MFCD10565758
CAS Number: 85774-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR