1-Benzylpiperidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A102422-10G A102422-1G A102422-250MG A102422-25G A102422-5G
76717-632EA 85 USD
76717-632 76717-634 76717-636 76717-638 76717-640
1-Benzylpiperidine-2-carboxylic acid 97%
1-Benzylpiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MDL Number: MFCD00756921
CAS Number: 21319-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR