N-Me-Val-OMe·HCl 95%

Supplier: Ambeed
Warning

Synonyms: N-Me-Val-OMe HCl, N-Methyl-L-valine methyl ester hydrochloride, N-Me-Val-OMe·HCl

A102416-250MG A102416-1G A102416-5G
76717-630EA 14 USD
76717-630 77548-932 77570-128
N-Me-Val-OMe·HCl 95%
N-Me-Val-OMe·HCl

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
MW: 181.66 g/mol
MDL Number: MFCD00238325
CAS Number: 3339-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR