Methyl thiazole-4-carboxylate 98%

Supplier: Ambeed
Warning

A101968-1G A101968-25G A101968-5G
76717-610EA 9 USD
76717-610 76717-612 76717-614
Methyl thiazole-4-carboxylate 98%
Methyl thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅NO₂S
MDL Number: MFCD06410940
CAS Number: 59418-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR