Mosapride citrate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-chloro-2-ethoxy-N-((4-(4-fluorobenzyl)morpholin-2-yl)methyl)benzamide 2-hydroxypropane-1,2,3-tricarboxylate, TAK-370, AS-4370 citrate

A101355-10MG A101355-1MG A101355-50MG
76717-526EA 10.61 USD
76717-526 76717-528 76717-530
Mosapride citrate 98%
Mosapride citrate

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClFN₃O₃·C₆H₈O₇
MW: 614.02 g/mol
MDL Number: MFCD01666680
CAS Number: 112885-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR