5-(Trifluoromethyl)dibenzothiophenium tetrafluoroborate 95%

Supplier: Ambeed
Warning

A100642-100MG A100642-1G A100642-250MG A100642-5G
76717-494EA 22 USD
76717-494 76717-496 76717-498 76717-500
5-(Trifluoromethyl)dibenzothiophenium tetrafluoroborate 95%
5-(Trifluoromethyl)dibenzothiophenium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₃H₈BF₇S
MDL Number: MFCD01073545
CAS Number: 131880-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR