(1-Propyl)triphenylphosphonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: Triphenyl(propyl)phosphonium bromide, Triphenylpropylphosphonium bromide

A100228-1000G A100228-100G A100228-25G A100228-500G
76717-474EA 186.72 USD
76717-474 76717-476 76717-478 76717-480
(1-Propyl)triphenylphosphonium bromide 98%
(1-Propyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₁H₂₂BrP
MW: 385.28 g/mol
MDL Number: MFCD00011843
CAS Number: 6228-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR