(±)-1,2,3,4-Tetrahydro-2-naphthol 95%

Supplier: Ambeed
Warning

A999720-100MG A999720-1G A999720-250MG A999720-5G
76698-954EA 11 USD
76698-954 76698-956 76698-958 76698-960
(±)-1,2,3,4-Tetrahydro-2-naphthol 95%
(±)-1,2,3,4-Tetrahydro-2-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MDL Number: MFCD00045575
CAS Number: 530-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR