Copper(II) oxide 99%
Supplier: Ambeed
|
Danger
|
A989426-25G
A989426-500G
A989426-1000G
A989426-100G
76698-948EA
11
USD
76698-948
77569-716
77603-402
77880-803
Copper(II) oxide 99%
Copper(II) oxide
Precaución: For research use only.
Formula:
CuO |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...