(R)-(+)-α-Ethylbenzylamine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-1-Phenylpropan-1-amine, (R)-(+)-1-Phenylpropylamine

A986339-1G A986339-25G A986339-5G
76698-940EA 9.55 USD
76698-940 76698-942 77573-946
(R)-(+)-α-Ethylbenzylamine 97%
(R)-(+)-α-Ethylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00083057
CAS Number: 3082-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR