(Chloromethyl)dimethylphenylsilane 97%

Supplier: Ambeed
Warning

Synonyms: (Chloromethyl)dimethyl(phenyl)silane

A985316-1G A985316-250MG A985316-25G A985316-5G
76698-932EA 15 USD
76698-932 76698-934 76698-936 76698-938
(Chloromethyl)dimethylphenylsilane 97%
(Chloromethyl)dimethylphenylsilane

Precaución: for Research Use Only

Formula: C₉H₁₃ClSi
MW: 184.7400 g/mol
MDL Number: MFCD00040858
CAS Number: 1833-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR