3-Chlorophenethyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 3-Chlorophenethyl Bromide

A985220-10G A985220-1G A985220-5G A985220-25G
76698-926EA 85 USD
76698-926 76698-928 76698-930 77878-776
3-Chlorophenethyl bromide 97%
3-Chlorophenethyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrCl
MW: 219.51 g/mol
MDL Number: MFCD04117465
CAS Number: 16799-05-6
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR