Methyl 2-amino-2-iminoacetate hydrochloride 95%

Supplier: Ambeed
Warning

A983270-100MG A983270-1G A983270-250MG A983270-5G A983270-25G
76698-914EA 22 USD
76698-914 76698-916 76698-918 77595-576 77627-792
Methyl 2-amino-2-iminoacetate hydrochloride 95%
Methyl 2-amino-2-iminoacetate hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇ClN₂O₂
MW: 138.55 g/mol
MDL Number: MFCD09608055
CAS Number: 60189-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR