Demethoxycurcumin 98%

Supplier: Ambeed
Warning

Synonyms: (1E,6E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)hepta-1,6-diene-3,5-dione

A982362-100MG A982362-250MG A982362-50MG
76698-912EA 222 USD
76698-912 77627-790 77880-795
Demethoxycurcumin 98%
Demethoxycurcumin

Precaución: for Research Use Only

Formula: C₂₀H₁₈O₅
MDL Number: MFCD03427310
CAS Number: 22608-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR