Methylene Violet 3RAX 98%

Supplier: Ambeed
Warning

A981648-1G A981648-25G A981648-5G
76698-906EA 22 USD
76698-906 76698-908 76698-910
Methylene Violet 3RAX 98%
Methylene Violet 3RAX

Precaución: for Research Use Only

Formula: C₂₂H₂₃ClN₄
MDL Number: MFCD00013433
CAS Number: 4569-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR