5-Bromo-2-hydroxy-3-methylbenzaldehyde 95%

Supplier: Ambeed
Warning

A980401-1G A980401-250MG A980401-5G A980401-25G
76698-902EA 27 USD
76698-902 76698-904 77603-382 77878-770
5-Bromo-2-hydroxy-3-methylbenzaldehyde 95%
5-Bromo-2-hydroxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MDL Number: MFCD01040324
CAS Number: 33172-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR