Boc-Cys(Et)-OH 97%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(ethylthio)propanoic acid

A979081-5G A979081-1G
76698-900EA 99 USD
76698-900 77878-769
Boc-Cys(Et)-OH 97%
Boc-Cys(Et)-OH

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄S
MW: 249.325 g/mol
MDL Number: MFCD00076919
CAS Number: 16947-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR