4,4-Dimethoxybutanone 90%

Supplier: Ambeed
Danger

Synonyms: 4,4-Dimethoxybutan-2-one, 3-Oxobutyraldehyde dimethyl acetal, Acetoacetaldehyde-1,1-Dimethyl Acetal, Acetylacetaldehyde dimethyl acetal, 4,4-Dimethoxy-2-butanone

A973773-10G A973773-25G A973773-100G
76698-886EA 5.31 USD
76698-886 77552-800 77603-374
4,4-Dimethoxybutanone 90%
4,4-Dimethoxybutanone

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
MDL Number: MFCD00008789
CAS Number: 5436-21-5
UN: 1989
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR