5-Bromo-2-fluorocinnamic acid 98%

Supplier: Ambeed
Warning

A971981-10G A971981-1G A971981-25G
76698-880EA 32.89 USD
76698-880 76698-882 76698-884
5-Bromo-2-fluorocinnamic acid 98%
5-Bromo-2-fluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
MDL Number: MFCD00143427
CAS Number: 202865-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR