EDTA calcium disodium salt 98+%

Supplier: Ambeed
Warning

Synonyms: Ethylenediaminetetraacetic acid disodium calcium salt, Calcium disodium ethylenediaminetetraacetate, Ethylenediaminetetraacetic acid calcium disodium salt hydrate, Ethylenedinitrilotetraacetic acid calcium disodium salt, Ethylenediaminetetraacetic Acid Calcium Disodium Salt

A968307-100G A968307-25G A968307-500G
76698-872EA 10 USD
76698-872 76698-874 76698-876
EDTA calcium disodium salt 98+%
EDTA calcium disodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₂CaN₂Na₂O₈
MW: 374.2684 g/mol
MDL Number: MFCD00148919
CAS Number: 62-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR