3-(Boc-amino)benzonitrile 98%

Supplier: Ambeed
Warning

A967003-250MG A967003-5G A967003-1G
76698-868EA 10 USD
76698-868 76698-870 77878-764
3-(Boc-amino)benzonitrile 98%
3-(Boc-amino)benzonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD07787223
CAS Number: 145878-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR