3-(Cyclopropylcarbamoyl)-5-nitrophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-(Cyclopropylcarbamoyl)-5-nitrophenyl)boronic acid

A967001-1G A967001-250MG A967001-5G
76698-862EA 78.51 USD
76698-862 76698-864 76698-866
3-(Cyclopropylcarbamoyl)-5-nitrophenylboronic acid 98%
3-(Cyclopropylcarbamoyl)-5-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BN₂O₅
MW: 250.0200 g/mol
MDL Number: MFCD07783878
CAS Number: 871332-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR