Diazene-1,2-diylbis(morpholinomethanone) 97%

Supplier: Ambeed
Danger

Synonyms: Azodicarboxylic bismorpholide, Azodicarboxylic dimorpholide

A965599-25G A965599-5G A965599-1G
76698-858EA 169.75 USD
76698-858 76698-860 77627-786
Diazene-1,2-diylbis(morpholinomethanone) 97%
Diazene-1,2-diylbis(morpholinomethanone)

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₄O₄
MW: 256.25 g/mol
MDL Number: MFCD00043006
CAS Number: 10465-82-4
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR