Bis(cyclohexanone)oxalyldihydrazone ≥99% (by HPLC) for spectrophotometric determination of Cu

Supplier: Ambeed
Danger

Synonyms: Cupferazone, N'1,N'2-Dicyclohexylideneoxalohydrazide, Oxalic acid bis(cyclohexylidenehydrazide), Bis(cyclohexanone)oxaldihydrazone, Cuprizone

A965540-100G A965540-25G A965540-500G
76698-852EA 29 USD
76698-852 76698-854 76698-856
Bis(cyclohexanone)oxalyldihydrazone ≥99% (by HPLC) for spectrophotometric determination of Cu
Bis(cyclohexanone)oxalyldihydrazone

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₄O₂
MW: 278.36 g/mol
MDL Number: MFCD00001659
CAS Number: 370-81-0
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR