Boc-L-Tic(7-OH)-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(tert-Butoxycarbonyl)-7-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-L-7-hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A956898-100MG A956898-1G A956898-250MG
76698-840EA 68 USD
76698-840 76698-842 76698-844
Boc-L-Tic(7-OH)-OH 95%
Boc-L-Tic(7-OH)-OH

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₅
MW: 293.319 g/mol
MDL Number: MFCD00671683
CAS Number: 142335-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR