3',5'-Difluoropropiophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Difluorophenyl)propan-1-one

A951202-10G A951202-1G
76698-818EA 94.43 USD
76698-818 76698-820
3',5'-Difluoropropiophenone 97%
3',5'-Difluoropropiophenone

Precaución: for Research Use Only

Formula: C₉H₈F₂O
MW: 170.16 g/mol
MDL Number: MFCD00061142
CAS Number: 135306-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR